Ethyl-2-aminobenzo[d]thiazole-6-carboxylate 98%

Supplier: Apollo Scientific
Warning

Synonyms: Ethyl 2-aminobenzo[d]thiazole-6-carboxylate

OR27460-25G OR27460-5G
APOSOR27460-25GEA 200 EUR
APOSOR27460-25G APOSOR27460-5G
Ethyl-2-aminobenzo[d]thiazole-6-carboxylate 98%
Ethyl-2-aminobenzo[d]thiazole-6-carboxylate
Formula: C₁₀H₁₀N₂O₂S
MW: 222,26 g/mol
Melting Pt: 241…243 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00102724
CAS Number: 50850-93-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR