2-Iodo-3-nitrobenzoic acid
Supplier: Apollo Scientific
|
Warning
|
OR27519-1G
OR27519-25G
OR27519-5G
APOSOR27519-1GEA
17.6
EUR
APOSOR27519-1G
APOSOR27519-25G
APOSOR27519-5G
2-Iodo-3-nitrobenzoic acid
2-Iodo-3-nitrobenzoic acid
Formula:
C₇H₄INO₄ MW: 293,02 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00277941 CAS Number: 5398-69-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...