3-(6-Methoxy-3-nitropyridin-2-yl)-2-methylpropanoic acid

Supplier: Apollo Scientific

OR27521-1G
APOSOR27521-1GEA 94.4 EUR
APOSOR27521-1G
3-(6-Methoxy-3-nitropyridin-2-yl)-2-methylpropanoic acid
3-(6-Methoxy-3-nitropyridin-2-yl)-2-methylpropanoic acid
Formula: C₁₀H₁₂N₂O₅
MW: 240,22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12827862
CAS Number: 175205-01-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR