4-Chlorooxindole
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
4-Chloroindolin-2-one
OR2754-25G
OR2754-5G
APOSOR2754-25GEA
279
EUR
APOSOR2754-25G
APOSOR2754-5G
4-Chlorooxindole
4-Chlorooxindole
Formula:
C₈H₆ClNO MW: 167,5900 g/mol Melting Pt: 206…208 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03787567 CAS Number: 20870-77-3 |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...