3,5-Bis(methylthio)isothiazole-4-carbonitrile 95+%
Supplier: Apollo Scientific
OR2763-25G
OR2763-5G
OR2763-1G
APOSOR2763-25GEA
1200
EUR
APOSOR2763-25G
APOSOR2763-5G
APOSOR2763-1G
3,5-Bis(methylthio)isothiazole-4-carbonitrile 95+%
3,5-Bis(methylthio)isothiazole-4-carbonitrile
Formula:
C₆H₆N₂S₃ Melting Pt: 130…132 °C Storage Temperature: Ambient |
MDL Number:
MFCD00173834 CAS Number: 4886-13-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...