Cyclododecanone oxime
Supplier: Apollo Scientific
|
Warning
|
OR27687-1G
OR27687-500MG
APOSOR27687-1GEA
230
EUR
APOSOR27687-1G
APOSOR27687-500MG
Cyclododecanone oxime
Cyclododecanone oxime
Formula:
C₁₂H₂₃NO Storage Temperature: Ambient |
CAS Number:
946-89-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...