4'-Methoxybutyrophenone

Supplier: Apollo Scientific

Synonyms: 1-(4-Methoxyphenyl)butan-1-one

OR27758-1G OR27758-5G
APOSOR27758-1GEA 56.8 EUR
APOSOR27758-1G APOSOR27758-5G
4'-Methoxybutyrophenone
4'-Methoxybutyrophenone
Formula: C₁₁H₁₄O₂
MW: 178,23 g/mol
Boiling Pt: 124…128 °C/4mmHg
Storage Temperature: Ambient
MDL Number: MFCD00027138
CAS Number: 4160-51-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR