7-(Diethylamino)coumarin-3-carboxylic acid
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
7ACC1, 7-(Diethylamino)-2-oxo-2H-chromene-3-carboxylic acid, DEAC, acid
OR27766-1G
OR27766-5G
APOSOR27766-1GEA
66.9
EUR
APOSOR27766-1G
APOSOR27766-5G
7-(Diethylamino)coumarin-3-carboxylic acid
7-(Diethylamino)coumarin-3-carboxylic acid
Formula:
C₁₄H₁₅NO₄ MW: 261,28 g/mol Melting Pt: 222…224 °C Storage Temperature: Ambient |
MDL Number:
MFCD00068721 CAS Number: 50995-74-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...