2-Amino-4'-bromoacetophenone hydrochloride
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
4-Bromophenacylamine hydrochloride
OR2785-1G
OR2785-250MG
OR2785-5G
APOSOR2785-1GEA
60.9
EUR
APOSOR2785-1G
APOSOR2785-250MG
APOSOR2785-5G
2-Amino-4'-bromoacetophenone hydrochloride
2-Amino-4'-bromoacetophenone hydrochloride
Formula:
C₈H₉BrClNO MW: 250,53 g/mol Melting Pt: 280…285 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00051998 CAS Number: 5467-72-1 EINECS: 226-778-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...