7-Azaindole 98%

Supplier: Apollo Scientific
Warning

Synonyms: 1H-Pyrrolo[2,3-b]pyridine

OR27924-5G OR27924-25G
APOSOR27924-5GEA 21.9 EUR
APOSOR27924-5G APOSOR27924-25G
7-Azaindole 98%
7-Azaindole
Formula: C₇H₆N₂
MW: 118,14 g/mol
Boiling Pt: 270 °C
Melting Pt: 103…110 °C
Storage Temperature: Ambient
MDL Number: MFCD00005606
CAS Number: 271-63-6
EINECS: 205-981-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR