Ethyl-3-hydroxy-2-naphthoate

Supplier: Apollo Scientific
Warning

Synonyms: 3-Hydroxy-2-naphthoic acid ethyl ester, Ethyl 3-hydroxy-2-naphthoate

OR27972-1G OR27972-250MG
APOSOR27972-1GEA 29 EUR
APOSOR27972-1G APOSOR27972-250MG
Ethyl-3-hydroxy-2-naphthoate
Ethyl 3-hydroxy-2-naphthoate
Formula: C₁₃H₁₂O₃
MW: 216,2326 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00220608
CAS Number: 7163-25-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR