Allylformamide

Supplier: Apollo Scientific

OR28268-1G OR28268-5G OR28268-10G
APOSOR28268-1GEA 78.3 EUR
APOSOR28268-1G APOSOR28268-5G APOSOR28268-10G
Allylformamide
Allylformamide
Formula: C₄H₇NO
MW: 85.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00964341
CAS Number: 16250-37-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR