4-Iodobenzenesulphonamide

Supplier: Apollo Scientific
Warning

OR2835-1G OR2835-5G
APOSOR2835-1GEA 21.9 EUR
APOSOR2835-1G APOSOR2835-5G
4-Iodobenzenesulphonamide
4-Iodobenzenesulphonamide
Formula: C₆H₆INO₂S
MW: 283.0900 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00092887
CAS Number: 825-86-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR