2-Bromo-5-methoxybenzoic acid 98%

Supplier: Apollo Scientific
Warning

OR28590-100G OR28590-500G
APOSOR28590-100GEA 101 EUR
APOSOR28590-100G APOSOR28590-500G
2-Bromo-5-methoxybenzoic acid 98%
2-Bromo-5-methoxybenzoic acid
Formula: C₈H₇BrO₃
MW: 231,05 g/mol
Melting Pt: 155…160 °C
Storage Temperature: Ambient
MDL Number: MFCD00020214
CAS Number: 22921-68-2
EINECS: 245-329-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR