Pyrrole 98%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
1H-Pyrrole
OR28598-1KG
OR28598-50G
OR28598-250G
APOSOR28598-1KGEA
603
EUR
APOSOR28598-1KG
APOSOR28598-50G
APOSOR28598-250G
Pyrrole 98%
Pyrrole
Formula:
C₄H₅N MW: 67.09 g/mol Boiling Pt: 129…131 °C Melting Pt: –23 °C Density: 0.96 g/cm³ (25 °C) Flash Pt: 36 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00005216 CAS Number: 109-97-7 EINECS: 203-724-7 UN: 1992 ADR: 3+6.1 III Merck Index: 12,08198 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...