4-Cyclohexylaniline
Supplier: Apollo Scientific
|
Warning
|
OR28600-1G
OR28600-10G
OR28600-5G
APOSOR28600-1GEA
36.4
EUR
APOSOR28600-1G
APOSOR28600-10G
APOSOR28600-5G
4-Cyclohexylaniline
4-Cyclohexylaniline
Formula:
C₁₂H₁₇N MW: 175.27 g/mol Boiling Pt: 166 °C/13mmHg Melting Pt: 52…56 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001454 CAS Number: 6373-50-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...