3,5-Dichloroaniline 98%
Supplier: Apollo Scientific
|
Danger
|
OR28608-1KG
OR28608-250G
OR28608-50G
APOSOR28608-1KGEA
422
EUR
APOSOR28608-1KG
APOSOR28608-250G
APOSOR28608-50G
3,5-Dichloroaniline 98%
3,5-Dichloroaniline
Formula:
C₆H₅Cl₂N MW: 162,02 g/mol Boiling Pt: 259…260 °C/741mmHg Melting Pt: 51…53 °C Density: 1,58 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00007774 CAS Number: 626-43-7 EINECS: 210-948-9 UN: 3442 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...