(3,4-Dimethoxyphenyl)acetic acid ethyl ester 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Ethyl (3,4-dimethoxyphenyl)acetate, Ethyl 2-(3,4-dimethoxyphenyl)acetate
OR28612-100G
OR28612-25G
APOSOR28612-100GEA
219
EUR
APOSOR28612-100G
APOSOR28612-25G
(3,4-Dimethoxyphenyl)acetic acid ethyl ester 98%
(3,4-Dimethoxyphenyl)acetic acid ethyl ester
Formula:
C₁₂H₁₆O₄ MW: 224,26 g/mol Boiling Pt: 159…160 °C/4mmHg Storage Temperature: Ambient |
MDL Number:
MFCD00017272 CAS Number: 18066-68-7 EINECS: 241-974-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...