Sodium triacetoxyborohydride 95%
Supplier: Apollo Scientific
|
Danger
|
OR2863-25G
OR2863-100G
OR2863-250G
APOSOR2863-25GEA
23.2
EUR
APOSOR2863-25G
APOSOR2863-100G
APOSOR2863-250G
Sodium triacetoxyborohydride 95%
Sodium triacetoxyborohydride
Formula:
C₆H₁₀BNaO₆ MW: 211,94 g/mol Melting Pt: 116…120 °C (dec.) Density: 1,43 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00012211 CAS Number: 56553-60-7 UN: 1409 ADR: 4.3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...