6-Methylanthranil acid 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-Amino-6-methylbenzoic acid
OR28634-100G
OR28634-25G
OR28634-5G
APOSOR28634-100GEA
230
EUR
APOSOR28634-100G
APOSOR28634-25G
APOSOR28634-5G
6-Methylanthranil acid 97%
6-Methylanthranil acid
Formula:
C₈H₉NO₂ Melting Pt: 125…128 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00007809 CAS Number: 4389-50-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...