2-Chloro-5-nitropyridine
Supplier: Apollo Scientific
|
Warning
|
OR28643-100G
OR28643-25G
OR28643-500G
APOSOR28643-100GEA
55.1
EUR
APOSOR28643-100G
APOSOR28643-25G
APOSOR28643-500G
2-Chloro-5-nitropyridine
2-Chloro-5-nitropyridine
Formula:
C₅H₃ClN₂O₂ MW: 158,54 g/mol Melting Pt: 105…108 °C Flash Pt: 158 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006240 CAS Number: 4548-45-2 EINECS: 224-908-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...