2,4-Dimethoxybenzaldehyde

Supplier: Apollo Scientific
Warning

OR28682-100G OR28682-500G OR28682-1KG
APOSOR28682-100GEA 79.9 EUR
APOSOR28682-100G APOSOR28682-500G APOSOR28682-1KG
2,4-Dimethoxybenzaldehyde
2,4-Dimethoxybenzaldehyde
Formula: C₉H₁₀O₃
MW: 166,18 g/mol
Boiling Pt: 165 °C/10mmHg
Melting Pt: 67…71 °C
Storage Temperature: Ambient
MDL Number: MFCD00003311
CAS Number: 613-45-6
EINECS: 210-342-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR