Diaminomaleonitrile 98%
Supplier: Apollo Scientific
|
Danger
|
OR28686-100G
OR28686-25G
APOSOR28686-100GEA
94.5
EUR
APOSOR28686-100G
APOSOR28686-25G
Diaminomaleonitrile 98%
Diaminomaleonitrile
Formula:
C₄H₄N₄ MW: 108,1 g/mol Melting Pt: 178…179 °C Density: 1,36 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00001870 CAS Number: 1187-42-4 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...