Phenyl isothiocyanate

Supplier: Apollo Scientific
Danger

OR28695-100G OR28695-500G
APOSOR28695-100GEA 88.7 EUR
APOSOR28695-100G APOSOR28695-500G
Phenyl isothiocyanate
Phenyl isothiocyanate
Formula: C₇H₅NS
Boiling Pt: 218…221 °C
Melting Pt: –21 °C
Density: 1,132 g/cm³ (25 °C)
Flash Pt: 87 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004798
CAS Number: 103-72-0
EINECS: 203-138-1
UN: 2927
ADR: 6.1+8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR