2-Phenyl-5-benzimidazolesulphonic acid

Supplier: Apollo Scientific
Danger

Synonyms: Ensulizole, Phenylbenzimidazole sulphonic acid, 1H-Benzimidazole-5-sulphonic acid

OR28712-25G OR28712-5G
APOSOR28712-25GEA 37.7 EUR
APOSOR28712-25G APOSOR28712-5G
2-Phenyl-5-benzimidazolesulphonic acid
2-Phenyl-5-benzimidazolesulphonic acid
Formula: C₁₃H₁₀N₂O₃S
MW: 274,29 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00053007
CAS Number: 27503-81-7
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR