3-Amino-1-phenyl-2-pyrazolin-5-one
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
5-Amino-2,4-dihydro-2-phenyl-3H-pyrazol-3-one
OR28715-10G
OR28715-1G
APOSOR28715-10GEA
62.6
EUR
APOSOR28715-10G
APOSOR28715-1G
3-Amino-1-phenyl-2-pyrazolin-5-one
3-Amino-1-phenyl-2-pyrazolin-5-one
Formula:
C₉H₉N₃O MW: 175,19 g/mol Melting Pt: 210…215 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003137 CAS Number: 4149-06-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...