Menadione (2-methyl-1,4-naphthoquinone, vitamin K3)

Supplier: Apollo Scientific
Warning

Synonyms: Menadione, 1,4-Dihydro-2-methylnaphthalene-1,4-dione, 2-Methylnaphthalene-1,4-dione

OR28733-25G OR28733-100G
APOSOR28733-25GEA 29 EUR
APOSOR28733-25G APOSOR28733-100G
Menadione (2-methyl-1,4-naphthoquinone, vitamin K3)
Menadione (2-Methyl-1,4-naphthoquinone, Vitamin K3)
Formula: C₁₁H₈O₂
MW: 172.19 g/mol
Melting Pt: 105…107 °C
Storage Temperature: Ambient
MDL Number: MFCD00001681
CAS Number: 58-27-5
EINECS: 200-372-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR