4-Bromoveratrole 98%

Supplier: Apollo Scientific

Synonyms: 1-Bromo-3,4-dimethoxybenzene, 4-Bromo-1,2-dimethoxybenzene

OR28742-500G
APOSOR28742-500GEA 246 EUR
APOSOR28742-500G
4-Bromoveratrole 98%
4-Bromoveratrole
Formula: C₈H₉BrO₂
MW: 217,06 g/mol
Boiling Pt: 256 °C
Density: 1,51 g/cm³ (25 °C)
Flash Pt: 109 °C
Storage Temperature: Ambient
MDL Number: MFCD00008381
CAS Number: 2859-78-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR