4'-Aminoacetophenone 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
1-(4-Aminophenyl)ethanone, p-Aminoacetophenone
OR28747-100G
OR28747-500G
APOSOR28747-100GEA
23.2
EUR
APOSOR28747-100G
APOSOR28747-500G
4'-Aminoacetophenone 98%
4'-Aminoacetophenone
Formula:
C₈H₉NO MW: 135,17 g/mol Boiling Pt: 292…294 °C Melting Pt: 103…107 °C Density: 0,77 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00007896 CAS Number: 99-92-3 EINECS: 202-801-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...