2-Chloroveratraldehyde

Supplier: Apollo Scientific
Warning

Synonyms: 2-Chloro-3,4-dimethoxybenzaldehyde

OR28781-25G
APOSOR28781-25GEA 72.5 EUR
APOSOR28781-25G
2-Chloroveratraldehyde
2-Chloroveratraldehyde
Formula: C₉H₉ClO₃
MW: 200,62 g/mol
Melting Pt: 70…74 °C
Storage Temperature: Ambient
MDL Number: MFCD00084942
CAS Number: 5417-17-4
EINECS: 226-515-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR