4-Acetamidobenzaldehyde 98%

Supplier: Apollo Scientific
Warning

Synonyms: N-(4-Formylphenyl)acetamide, 4'-Formylacetanilide

OR28813-25G OR28813-100G
APOSOR28813-25GEA 58 EUR
APOSOR28813-25G APOSOR28813-100G
4-Acetamidobenzaldehyde 98%
4-Acetamidobenzaldehyde
Formula: C₉H₉NO₂
MW: 163.18 g/mol
Melting Pt: 154…158 °C
Storage Temperature: Ambient
MDL Number: MFCD00003380
CAS Number: 122-85-0
EINECS: 204-579-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR