4,7-Dichloroquinoline
Supplier: Apollo Scientific
|
Warning
|
OR28817-100G
OR28817-25G
APOSOR28817-100GEA
46.6
EUR
APOSOR28817-100G
APOSOR28817-25G
4,7-Dichloroquinoline
4,7-Dichloroquinoline
Formula:
C₉H₅Cl₂N MW: 198,05 g/mol Boiling Pt: 148 °C/10mmHg Melting Pt: 81…87 °C Flash Pt: 164 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006774 CAS Number: 86-98-6 EINECS: 201-714-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...