3,4-Dihydroxy-3-cyclobutene-1,2-dione

Supplier: Apollo Scientific
Warning

Synonyms: Squaric acid, 1,2-Dihydroxy-3,4-cyclobutenedione, Quadratic acid, 1,2-Dihydroxy-1-cyclobutene-3,4-dione, Diketocyclobutenediol

OR28821-100G OR28821-25G OR28821-500G
APOSOR28821-100GEA 154 EUR
APOSOR28821-100G APOSOR28821-25G APOSOR28821-500G
3,4-Dihydroxy-3-cyclobutene-1,2-dione
3,4-Dihydroxy-3-cyclobutene-1,2-dione
Formula: C₄H₂O₄
MW: 114,06 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00001334
CAS Number: 2892-51-5
EINECS: 220-761-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR