1,3-Diphenyltriazene 95%
Supplier: Apollo Scientific
|
Warning
|
OR28839-1G
OR28839-500MG
OR28839-250MG
APOSOR28839-1GEA
560
EUR
APOSOR28839-1G
APOSOR28839-500MG
APOSOR28839-250MG
1,3-Diphenyltriazene 95%
1,3-Diphenyltriazene
Formula:
C₁₂H₁₁N₃ Boiling Pt: 146 °C Melting Pt: 98 °C Storage Temperature: Ambient |
CAS Number:
136-35-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...