2-Acetyl-6-methoxynaphthalene

Supplier: Apollo Scientific
Warning

Synonyms: 6'-Methoxy-2'-acetonaphthone

OR28862-100G OR28862-25G OR28862-500G
APOSOR28862-100GEA 24.8 EUR
APOSOR28862-100G APOSOR28862-25G APOSOR28862-500G
2-Acetyl-6-methoxynaphthalene
2-Acetyl-6-methoxynaphthalene
Formula: C₁₃H₁₂O₂
MW: 200,24 g/mol
Boiling Pt: 155…160 °C/0,4mmHg
Melting Pt: 107…109 °C
Storage Temperature: Ambient
MDL Number: MFCD00021643
CAS Number: 3900-45-6
EINECS: 223-453-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR