β-Chloropropiophenone 98%

Supplier: Apollo Scientific
Warning

Synonyms: 3-Chloropropiophenone

OR2891-100G OR2891-25G OR2891-500G
APOSOR2891-100GEA 43.7 EUR
APOSOR2891-100G APOSOR2891-25G APOSOR2891-500G
β-Chloropropiophenone 98%
β-Chloropropiophenone
Formula: (C₆H₅)C(O)CH₂CH₂Cl
MW: 168,62 g/mol
Boiling Pt: 113…115 °C/4mmHg
Melting Pt: 46…50 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00000990
CAS Number: 936-59-4
EINECS: 213-317-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR