2,5-Dihydro-2,5-dimethoxy-2-methylfuran

Supplier: Apollo Scientific
Warning

Synonyms: 2,5-Dimethoxy-2-methyl-2,5-dihydrofuran

OR28934-10G OR28934-1G OR28934-25G
APOSOR28934-10GEA 164 EUR
APOSOR28934-10G APOSOR28934-1G APOSOR28934-25G
2,5-Dihydro-2,5-dimethoxy-2-methylfuran
2,5-Dihydro-2,5-dimethoxy-2-methylfuran
Formula: C₇H₁₂O₃
MW: 144,1684 g/mol
Boiling Pt: 46 °C/10mmHg
Density: 1,019 g/cm³ (25 °C)
Flash Pt: 46 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00126939
CAS Number: 22414-24-0
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR