Isopropyl 2-bromo-2-methylpropionate 99%

Supplier: Apollo Scientific
Danger

OR28936-100G OR28936-25G
APOSOR28936-100GEA 58 EUR
APOSOR28936-100G APOSOR28936-25G
Isopropyl 2-bromo-2-methylpropionate 99%
Isopropyl 2-bromo-2-methylpropionate
Formula: C₇H₁₃BrO₂
Boiling Pt: 168…170 °C
Storage Temperature: Ambient
MDL Number: MFCD00055235
CAS Number: 51368-55-9
EINECS: 257-158-0
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR