2,3-Dihydrofuran 97%

Supplier: Apollo Scientific

OR2896-250G OR2896-500G
APOSOR2896-250GEA 139 EUR
APOSOR2896-250G APOSOR2896-500G
2,3-Dihydrofuran 97%
2,3-Dihydrofuran
Formula: C₄H₆O
MW: 70,09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003205
CAS Number: 1191-99-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR