Methyl-3-(5-formyl-2-furyl)-2-thiophenecarboxylate
Supplier: Apollo Scientific
OR29103-10G
OR29103-1G
OR29103-5G
APOSOR29103-10GEA
789
EUR
APOSOR29103-10G
APOSOR29103-1G
APOSOR29103-5G
Methyl-3-(5-formyl-2-furyl)-2-thiophenecarboxylate
Methyl-3-(5-formyl-2-furyl)-2-thiophenecarboxylate
Formula:
C₁₁H₈O₄S MW: 236,25 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00052169 CAS Number: 255828-29-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...