2-Chloro-5-ethylpyrimidine 97%

Supplier: Apollo Scientific
Warning

OR2912-100G OR2912-25G OR2912-5G
APOSOR2912-100GEA 231 EUR
APOSOR2912-100G APOSOR2912-25G APOSOR2912-5G
2-Chloro-5-ethylpyrimidine 97%
2-Chloro-5-ethylpyrimidine
Formula: C₆H₇ClN₂
MW: 142,59 g/mol
Boiling Pt: 223 °C
Density: 1,174 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00799503
CAS Number: 111196-81-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR