3,4-Dimethylthiophene

Supplier: Apollo Scientific

OR29155-1G OR29155-5G
APOSOR29155-1GEA 148 EUR
APOSOR29155-1G APOSOR29155-5G
3,4-Dimethylthiophene
3,4-Dimethylthiophene
Formula: C₆H₈S
MW: 112,1900 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01764731
CAS Number: 632-15-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR