6-Chloropiperonal

Supplier: Apollo Scientific
Warning

Synonyms: 6-Chloro-1,3-benzodioxole-5-carboxaldehyde

OR2927-1G OR2927-250MG
APOSOR2927-1GEA 290 EUR
APOSOR2927-1G APOSOR2927-250MG
6-Chloropiperonal
6-Chloropiperonal
Formula: C₈H₅ClO₃
MW: 184,58 g/mol
Melting Pt: 114…116 °C
Storage Temperature: Ambient
MDL Number: MFCD00022953
CAS Number: 15952-61-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR