Piperonylonitrile 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
3,4-(Methylenedioxy)benzonitrile, 1,3-Benzodioxole-5-carbonitrile
OR29275-25G
OR29275-1G
OR29275-5G
OR29275-10G
OR29275-50G
APOSOR29275-25GEA
102
EUR
APOSOR29275-25G
APOSOR29275-1G
APOSOR29275-5G
APOSOR29275-10G
APOSOR29275-50G
Piperonylonitrile 97%
Piperonylonitrile
Formula:
C₈H₅NO₂ MW: 147,13 g/mol Melting Pt: 90…94 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005820 CAS Number: 4421-09-4 EINECS: 224-590-6 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...