Ethyl-1-(2-formylphenyl)piperidine-4-carboxylate

Supplier: Apollo Scientific
Warning

Synonyms: Ethyl 1-(2-formylphenyl)piperidine-4-carboxylate

OR29347-1G OR29347-250MG
APOSOR29347-1GEA 102 EUR
APOSOR29347-1G APOSOR29347-250MG
Ethyl-1-(2-formylphenyl)piperidine-4-carboxylate
Ethyl-1-(2-formylphenyl)piperidine-4-carboxylate
Formula: C₁₅H₁₉NO₃
MW: 261,3200 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00662563
CAS Number: 259683-56-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR