2,2,2-Trichloro-1-(1H-pyrrol-2-yl)ethanone

Supplier: Apollo Scientific
Warning

Synonyms: 2-(Trichloroacetyl)pyrrole, 2-(trichloroacetyl)-1h-pyrrole

OR29374-100G OR29374-25G OR29374-500G
APOSOR29374-100GEA 87.3 EUR
APOSOR29374-100G APOSOR29374-25G APOSOR29374-500G
2,2,2-Trichloro-1-(1H-pyrrol-2-yl)ethanone
2,2,2-Trichloro-1-(1H-pyrrol-2-yl)ethanone
Formula: C₆H₄Cl₃NO
MW: 212,46 g/mol
Melting Pt: 72…74 °C
Storage Temperature: Ambient
MDL Number: MFCD00128757
CAS Number: 35302-72-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR