4-Bromo-2-(trichloroacetyl)pyrrole

Supplier: Apollo Scientific
Warning

Synonyms: 1-(4-Bromo-1H-pyrrol-2-yl)-2,2,2-trichloroethanone

OR29377-1G OR29377-5G
APOSOR29377-1GEA 85.8 EUR
APOSOR29377-1G APOSOR29377-5G
4-Bromo-2-(trichloroacetyl)pyrrole
4-Bromo-2-(trichloroacetyl)pyrrole
Formula: C₆H₃BrCl₃NO
MW: 291,3571 g/mol
Melting Pt: 134…136 °C
Storage Temperature: Ambient
MDL Number: MFCD00832836
CAS Number: 72652-32-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR