(2-Chlorophenoxy)acetonitrile

Supplier: Apollo Scientific

OR29618-10G OR29618-1G OR29618-5G
APOSOR29618-10GEA 80 EUR
APOSOR29618-10G APOSOR29618-1G APOSOR29618-5G
(2-Chlorophenoxy)acetonitrile
(2-Chlorophenoxy)acetonitrile
Formula: C₈H₆ClNO
MW: 167,60 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00017331
CAS Number: 43111-31-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR