6-Aminopyridazine-3-carboxylic acid

Supplier: Apollo Scientific
Warning

OR2974-1G OR2974-5G
APOSOR2974-1GEA 105 EUR
APOSOR2974-1G APOSOR2974-5G
6-Aminopyridazine-3-carboxylic acid
6-Aminopyridazine-3-carboxylic acid
Formula: C₅H₅N₃O₂
Storage Temperature: Ambient
MDL Number: MFCD06658328
CAS Number: 59772-58-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR