2-Chloro-4'-methoxyacetanilide
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-Chloro-N-(4-methoxyphenyl)acetamide
OR29760-1G
APOSOR29760-1GEA
50.9
EUR
APOSOR29760-1G
2-Chloro-4'-methoxyacetanilide
2-Chloro-4'-methoxyacetanilide
Formula:
C₉H₁₀ClNO₂ Melting Pt: 121…122 °C Storage Temperature: Ambient |
MDL Number:
MFCD00018907 CAS Number: 22303-36-2 |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...